4-Iodopyrimidin-5-amine
Lieferant: Apollo Scientific
OR345719-1G
APOSOR345719-1GEA
243
EUR
APOSOR345719-1G
4-Iodopyrimidin-5-amine
4-Iodopyrimidin-5-amine
Formel:
C₄H₄IN₃ Molecular Weight: 220,999 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17169943 CAS-Nummer: 942067-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...