1,2-Thiazinan-3-one 1,1-dioxide 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR3458-100MG
APOSOR3458-100MGEA
63.8
EUR
APOSOR3458-100MG
1,2-Thiazinan-3-one 1,1-dioxide 97%
1,2-Thiazinan-3-one 1,1-dioxide
Formel:
C₄H₇NO₃S Schmelzpunkt: 150…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689509 CAS-Nummer: 913836-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...