1-tert-Butyloxycarbonyl-4-(3-aminopropyl)piperazine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 4-(3-aminopropyl)piperazine-1-carboxylate
OR346019-5G
APOSOR346019-5GEA
215
EUR
APOSOR346019-5G
1-tert-Butyloxycarbonyl-4-(3-aminopropyl)piperazine 95+%
1-tert-Butyloxycarbonyl-4-(3-aminopropyl)piperazine
Formel:
C₁₂H₂₅N₃O₂ Molecular Weight: 243.35 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06808578 CAS-Nummer: 373608-48-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...