Ethyl-6-chloro-5-cyano-2-methylnicotinate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 6-chloro-5-cyano-2-methylpyridine-3-carboxylate
OR346021-1G
APOSOR346021-1GEA
0
EUR
APOSOR346021-1G
Ethyl-6-chloro-5-cyano-2-methylnicotinate
Ethyl-6-chloro-5-cyano-2-methylnicotinate
Formel:
C₁₀H₉ClN₂O₂ Schmelzpunkt: 66…68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00179894 CAS-Nummer: 64119-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...