Ethyl 6-chloro-5-cyano-2-phenylnicotinate

Lieferant: Apollo Scientific

OR346022-1G
APOSOR346022-1GEA 0 EUR
APOSOR346022-1G
Ethyl 6-chloro-5-cyano-2-phenylnicotinate
Ethyl 6-chloro-5-cyano-2-phenylnicotinate
Formel: C₁₅H₁₁ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01315278
CAS-Nummer: 303146-96-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR