Ethyl-5-(tert-butyloxycarbonylaminomethyl)[1,3,4]oxadiazole-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 5-(((tert-butoxycarbonyl)amino)methyl)-1,3,4-oxadiazole-2-carboxylate

OR346028-1G
APOSOR346028-1GEA 0 EUR
APOSOR346028-1G
Ethyl-5-(tert-butyloxycarbonylaminomethyl)[1,3,4]oxadiazole-2-carboxylate
Ethyl-5-(tert-butyloxycarbonylaminomethyl)[1,3,4]oxadiazole-2-carboxylate
Formel: C₁₁H₁₇N₃O₅
Molecular Weight: 271,28 g/mol
Schmelzpunkt: 66…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10568221
CAS-Nummer: 164029-51-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR