5,7-Dibromo-1,3-dihydroindol-2-one

Lieferant: Apollo Scientific

OR346030-1G
APOSOR346030-1GEA 0 EUR
APOSOR346030-1G
5,7-Dibromo-1,3-dihydroindol-2-one
5,7-Dibromo-1,3-dihydroindol-2-one
Formel: C₈H₅Br₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD04230619
CAS-Nummer: 23872-19-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR