2-Phenylthioacetamide
Lieferant: Apollo Scientific
OR346049-5G
APOSOR346049-5GEA
0
EUR
APOSOR346049-5G
2-Phenylthioacetamide
2-Phenylthioacetamide
Formel:
C₈H₉NS Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022177 CAS-Nummer: 645-54-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...