Ethyl 2-Benzylthiazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

OR346050-1G
APOSOR346050-1GEA 383 EUR
APOSOR346050-1G
Ethyl 2-Benzylthiazole-4-carboxylate
Ethyl 2-Benzylthiazole-4-carboxylate
Formel: C₁₃H₁₃NO₂S
Molecular Weight: 247,313 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02102511
CAS-Nummer: 83553-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR