2-(2-Bromophenyl)thioacetamide

Lieferant: Apollo Scientific

OR346052-1G
APOSOR346052-1GEA 0 EUR
APOSOR346052-1G
2-(2-Bromophenyl)thioacetamide
2-(2-Bromophenyl)thioacetamide
Formel: C₈H₈BrNS
Lagertemperatur: Raumtemperatur
MDL: MFCD09816252
CAS-Nummer: 103317-32-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR