Ethyl 5-(2-bromoacetyl)-3-methylisoxazole-4-carboxylate

Lieferant: Apollo Scientific

OR346063-1G
APOSOR346063-1GEA 0 EUR
APOSOR346063-1G
Ethyl 5-(2-bromoacetyl)-3-methylisoxazole-4-carboxylate
Ethyl 5-(2-bromoacetyl)-3-methylisoxazole-4-carboxylate
Formel: C₉H₁₀BrNO₄
Molecular Weight: 276,084 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11052381
CAS-Nummer: 129663-20-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR