2-Phenylacetamidoxime

Lieferant: Apollo Scientific
Achtung

OR346070-5G
APOSOR346070-5GEA 139 EUR
APOSOR346070-5G
2-Phenylacetamidoxime
2-Phenylacetamidoxime
Formel: C₈H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00019953
CAS-Nummer: 19227-11-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR