2-(4-Bromophenyl)-N'-hydroxyethanimidamide

Lieferant: Apollo Scientific

Synonyme: 2-(4-Bromophenyl)-N-hydroxyacetimidamide, 2-(4-Bromophenyl)acetamidoxime

OR346071-1G
APOSOR346071-1GEA 0 EUR
APOSOR346071-1G
2-(4-Bromophenyl)-N'-hydroxyethanimidamide
2-(4-Bromophenyl)-N'-hydroxyethanimidamide
Formel: C₈H₉BrN₂O
Molecular Weight: 229,077 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05662829
CAS-Nummer: 422560-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR