5-(tert-Butyloxycarbonyamino)methyl-3-pyridin-3-yl[1,2,4]oxadiazole
Lieferant: Apollo Scientific
OR346076-1G
APOSOR346076-1GEA
0
EUR
APOSOR346076-1G
5-(tert-Butyloxycarbonyamino)methyl-3-pyridin-3-yl[1,2,4]oxadiazole
5-(tert-Butyloxycarbonyamino)methyl-3-pyridin-3-yl[1,2,4]oxadiazole
Formel:
C₁₃H₁₆N₄O₃ Molecular Weight: 276,3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10568315 CAS-Nummer: 1053656-62-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...