[2-(2-Bromobenzyl)thiazol-4-ylacetic acid

Lieferant: Apollo Scientific

OR346078-1G
APOSOR346078-1GEA 0 EUR
APOSOR346078-1G
[2-(2-Bromobenzyl)thiazol-4-ylacetic acid
[2-(2-Bromobenzyl)thiazol-4-ylacetic acid
Formel: C₁₂H₁₀BrNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD10568321
CAS-Nummer: 1053656-88-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR