[2-(4-Bromobenzyl)thiazol-4-y]acetic acid
Lieferant: Apollo Scientific
OR346079-1G
APOSOR346079-1GEA
0
EUR
APOSOR346079-1G
[2-(4-Bromobenzyl)thiazol-4-y]acetic acid
[2-(4-Bromobenzyl)thiazol-4-y]acetic acid
Formel:
C₁₂H₁₀BrNO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD10568322 CAS-Nummer: 1053656-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...