5-Chloromethyl-2-(4-chlorophenyl)-1H-imidazole

Lieferant: Apollo Scientific

OR346083-1G
APOSOR346083-1GEA 0 EUR
APOSOR346083-1G
5-Chloromethyl-2-(4-chlorophenyl)-1H-imidazole
5-Chloromethyl-2-(4-chlorophenyl)-1H-imidazole
Formel: C₁₀H₈Cl₂N₂
Molecular Weight: 227,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10568334
CAS-Nummer: 1053657-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR