2-(5-Chloromethyl-1H-imidazol-2-yl)pyridine

Lieferant: Apollo Scientific

OR346084-1G
APOSOR346084-1GEA 0 EUR
APOSOR346084-1G
2-(5-Chloromethyl-1H-imidazol-2-yl)pyridine
2-(5-Chloromethyl-1H-imidazol-2-yl)pyridine
Formel: C₉H₈ClN₃
Molecular Weight: 193,633 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10568335
CAS-Nummer: 1053657-25-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR