Ethyl-5-acetyl-2-methylthiazole-4-carboxylate

Lieferant: Apollo Scientific

OR346086-1G
APOSOR346086-1GEA 0 EUR
APOSOR346086-1G
Ethyl-5-acetyl-2-methylthiazole-4-carboxylate
Ethyl-5-acetyl-2-methylthiazole-4-carboxylate
Formel: C₉H₁₁NO₃S
Molecular Weight: 213,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173821
CAS-Nummer: 175277-29-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR