Ethyl-5-bromomethyl-3-methylisoxazole-4-carboxylate

Lieferant: Apollo Scientific

OR346091-1G
APOSOR346091-1GEA 0 EUR
APOSOR346091-1G
Ethyl-5-bromomethyl-3-methylisoxazole-4-carboxylate
Ethyl-5-bromomethyl-3-methylisoxazole-4-carboxylate
Formel: C₈H₁₀BrNO₃
Molecular Weight: 248,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10568349
CAS-Nummer: 159770-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR