Ethyl-5-formyl-3-methylisoxazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

OR346092-1G
APOSOR346092-1GEA 0 EUR
APOSOR346092-1G
Ethyl-5-formyl-3-methylisoxazole-4-carboxylate
Ethyl-5-formyl-3-methylisoxazole-4-carboxylate
Formel: C₈H₉NO₄
Molecular Weight: 183,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10568350
CAS-Nummer: 129663-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR