Methyl-2-benzyloxycarbonylamino-3-hydroxypropionate
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-(((benzyloxy)carbonyl)amino)-3-hydroxypropanoate
OR346093-25G
APOSOR346093-25GEA
173
EUR
APOSOR346093-25G
Methyl-2-benzyloxycarbonylamino-3-hydroxypropionate
Methyl-2-benzyloxycarbonylamino-3-hydroxypropionate
Formel:
C₁₂H₁₅NO₅ Molecular Weight: 253,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00136653 CAS-Nummer: 14464-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...