tert-Butyl (N-hydroxycarbamimidoylmethyl)carbamate

Lieferant: Apollo Scientific

OR346094-1G
APOSOR346094-1GEA 0 EUR
APOSOR346094-1G
tert-Butyl (N-hydroxycarbamimidoylmethyl)carbamate
tert-Butyl (N-hydroxycarbamimidoylmethyl)carbamate
Formel: C₇H₁₅N₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11052410
CAS-Nummer: 479079-15-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR