5-Bromo-2-chloromethyl-1H-benzoimidazole

Lieferant: Apollo Scientific

OR346101-1G OR346101-5G OR346101-10G
APOSOR346101-1GEA 0 EUR
APOSOR346101-1G APOSOR346101-5G APOSOR346101-10G
5-Bromo-2-chloromethyl-1H-benzoimidazole
5-Bromo-2-chloromethyl-1H-benzoimidazole
Formel: C₈H₆BrClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00457491
CAS-Nummer: 1740-88-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR