3-(4-Bromophenyl)-5-chloro[1,2,4]thiadiazole
Lieferant: Apollo Scientific
OR346103-1G
APOSOR346103-1GEA
0
EUR
APOSOR346103-1G
3-(4-Bromophenyl)-5-chloro[1,2,4]thiadiazole
3-(4-Bromophenyl)-5-chloro[1,2,4]thiadiazole
Formel:
C₈H₄BrClN₂S Molecular Weight: 275,56 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11052445 CAS-Nummer: 191919-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...