Methyl-6-methoxy-1H-indole-2-carboxylate
Lieferant: Apollo Scientific
Synonyme:
methyl 6-methoxyindole-2-carboxylate, Methyl 6-methoxy-1H-indole-2-carboxylate, Methyl 6-methoxy-2-indolecarboxylate, methyl 6-methoxy-1H-indole-2-carboxylate
OR346106-1G
APOSOR346106-1GEA
0
EUR
APOSOR346106-1G
Methyl-6-methoxy-1H-indole-2-carboxylate
Methyl-6-methoxy-1H-indole-2-carboxylate
Formel:
C₁₁H₁₁NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134301 CAS-Nummer: 98081-83-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...