4-Methyl-1H-indole-2-carboxylic acid methyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-Methylindole-2-carboxylate

OR346108-500MG
APOSOR346108-500MGEA 0 EUR
APOSOR346108-500MG
4-Methyl-1H-indole-2-carboxylic acid methyl ester
4-Methyl-1H-indole-2-carboxylic acid methyl ester
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189,22 g/mol
Schmelzpunkt: 117…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04966966
CAS-Nummer: 136831-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR