4,6-Dichloro-2-(methylthio)pyrimidine-5-carbaldehyde

Lieferant: Apollo Scientific

OR346115-25G
APOSOR346115-25GEA 190 EUR
APOSOR346115-25G
4,6-Dichloro-2-(methylthio)pyrimidine-5-carbaldehyde
4,6-Dichloro-2-(methylthio)pyrimidine-5-carbaldehyde
Formel: C₆H₄Cl₂N₂OS
Molecular Weight: 223.08 g/mol
Schmelzpunkt: 86…91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00194954
CAS-Nummer: 33097-11-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR