2-Chloro-3-chloromethyl-pyridine 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Chloro-3-(chloromethyl)pyridine

OR346119-5G
APOSOR346119-5GEA 105 EUR
APOSOR346119-5G
2-Chloro-3-chloromethyl-pyridine 98%
2-Chloro-3-chloromethyl-pyridine
Formel: C₆H₅Cl₂N
Molecular Weight: 162.017 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09835107
CAS-Nummer: 89581-84-0
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR