6-Acetyl-2-mercaptonicotinonitrile

Lieferant: Apollo Scientific

OR346127-1G
APOSOR346127-1GEA 0 EUR
APOSOR346127-1G
6-Acetyl-2-mercaptonicotinonitrile
6-Acetyl-2-mercaptonicotinonitrile
Formel: C₈H₆N₂OS
Molecular Weight: 178,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00204161
CAS-Nummer: 175277-42-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR