3-Chloro-5-iodopyridin-2-ol

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-5-iodo-pyridin-2-ol

OR346132-1G
APOSOR346132-1GEA 0 EUR
APOSOR346132-1G
3-Chloro-5-iodopyridin-2-ol
3-Chloro-5-iodopyridin-2-ol
Formel: C₅H₃ClINO
Molecular Weight: 255,44 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11227187
CAS-Nummer: 97966-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR