6-Bromomethyl-nicotinic acid methyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 6-(bromomethyl)nicotinate

OR346134-250MG
APOSOR346134-250MGEA 39.3 EUR
APOSOR346134-250MG
6-Bromomethyl-nicotinic acid methyl ester
6-Bromomethyl-nicotinic acid methyl ester
Formel: C₈H₈BrNO₂
Molecular Weight: 230.07 g/mol
Schmelzpunkt: 72…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10566280
CAS-Nummer: 131803-48-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR