(E)-2-Methyl-3-(1-phenylethylamino)but-2-enoic acid ethyl ester

Lieferant: Apollo Scientific

OR346135-1G
APOSOR346135-1GEA 0 EUR
APOSOR346135-1G
(E)-2-Methyl-3-(1-phenylethylamino)but-2-enoic acid ethyl ester
(E)-2-Methyl-3-(1-phenylethylamino)but-2-enoic acid ethyl ester
Formel: C₁₅H₂₁NO₂
Molecular Weight: 247,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11227202
CAS-Nummer: 92701-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR