3-Chloro-pyrazine-2-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-chloropyrazine-2-carboxylate

OR346138-5G
APOSOR346138-5GEA 75.4 EUR
APOSOR346138-5G
3-Chloro-pyrazine-2-carboxylic acid ethyl ester
3-Chloro-pyrazine-2-carboxylic acid ethyl ester
Formel: C₇H₇ClN₂O₂
Molecular Weight: 186.596 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371661
CAS-Nummer: 655247-45-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR