3-Methyl-isothiazole
Lieferant: Apollo Scientific
|
Achtung
|
OR346143-1G
APOSOR346143-1GEA
607
EUR
APOSOR346143-1G
3-Methyl-isothiazole
3-Methyl-isothiazole
Formel:
C₄H₅NS Molecular Weight: 99,154 g/mol Siedepunkt: 130…133 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00955579 CAS-Nummer: 693-92-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...