5-Methyl-2-phenyl-oxazole-4-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

OR346144-1G
APOSOR346144-1GEA 0 EUR
APOSOR346144-1G
5-Methyl-2-phenyl-oxazole-4-carboxylic acid ethyl ester
5-Methyl-2-phenyl-oxazole-4-carboxylic acid ethyl ester
Formel: C₁₃H₁₃NO₃
Molecular Weight: 231.247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05244510
CAS-Nummer: 61151-96-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR