5-Chloro-3-p-tolyl-[1,2,4]thiadiazole

Lieferant: Apollo Scientific
Achtung

OR346147-1G
APOSOR346147-1GEA 0 EUR
APOSOR346147-1G
5-Chloro-3-p-tolyl-[1,2,4]thiadiazole
5-Chloro-3-p-tolyl-[1,2,4]thiadiazole
Formel: C₉H₇ClN₂S
Molecular Weight: 210,683 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11227244
CAS-Nummer: 221038-01-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR