2-(2-Methyl-2H-pyrazol-3-yl)phenylamine

Lieferant: Apollo Scientific

OR346153-1G
APOSOR346153-1GEA 0 EUR
APOSOR346153-1G
2-(2-Methyl-2H-pyrazol-3-yl)phenylamine
2-(2-Methyl-2H-pyrazol-3-yl)phenylamine
Formel: C₁₀H₁₁N₃
Molecular Weight: 173,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845689
CAS-Nummer: 1208081-56-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR