3-Amino-4-methyl-benzo[b]thiophene-2-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

OR346155-1G
APOSOR346155-1GEA 0 EUR
APOSOR346155-1G
3-Amino-4-methyl-benzo[b]thiophene-2-carboxylic acid ethyl ester
3-Amino-4-methyl-benzo[b]thiophene-2-carboxylic acid ethyl ester
Formel: C₁₂H₁₃NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD11845694
CAS-Nummer: 181284-94-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR