6-Methylsulphanyl-benzothiazol-2-ylamine

Lieferant: Apollo Scientific

OR346157-1G
APOSOR346157-1GEA 0 EUR
APOSOR346157-1G
6-Methylsulphanyl-benzothiazol-2-ylamine
6-Methylsulphanyl-benzothiazol-2-ylamine
Formel: C₈H₈N₂S₂
Molecular Weight: 196.293 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845702
CAS-Nummer: 50850-92-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR