1-(4-Chlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid

Lieferant: Apollo Scientific

OR346165-1G OR346165-500MG
APOSOR346165-1GEA 0 EUR
APOSOR346165-1G APOSOR346165-500MG
1-(4-Chlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
1-(4-Chlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
Formel: C₁₂H₁₁ClN₂O₂
Molecular Weight: 250.69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09741134
CAS-Nummer: 113808-87-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR