6-tert-Butyl-nicotinonitrile

Lieferant: Apollo Scientific

OR346171-1G
APOSOR346171-1GEA 0 EUR
APOSOR346171-1G
6-tert-Butyl-nicotinonitrile
6-tert-Butyl-nicotinonitrile
Formel: C₁₀H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11845758
CAS-Nummer: 56029-45-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR