1-Methyl-1H-pyrazole-3,5-dicarboxylic acid dimethyl ester

Lieferant: Apollo Scientific

Synonyme: Dimethyl 1-methyl-1H-pyrazole-3,5-dicarboxylate

OR346173-5G
APOSOR346173-5GEA 101 EUR
APOSOR346173-5G
1-Methyl-1H-pyrazole-3,5-dicarboxylic acid dimethyl ester
1-Methyl-1H-pyrazole-3,5-dicarboxylic acid dimethyl ester
Formel: C₈H₁₀N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD08443806
CAS-Nummer: 33146-99-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR