6-Bromobenzo[b]thiophene-2-carbaldehyde

Lieferant: Apollo Scientific

OR346188-1G
APOSOR346188-1GEA 0 EUR
APOSOR346188-1G
6-Bromobenzo[b]thiophene-2-carbaldehyde
6-Bromobenzo[b]thiophene-2-carbaldehyde
Formel: C₉H₅BrOS
Lagertemperatur: Raumtemperatur
MDL: MFCD09834665
CAS-Nummer: 19075-45-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR