5-Bromo-benzo[b]thiophene-2-carbaldehyde

Lieferant: Apollo Scientific

Synonyme: 5-Bromobenzo[b]thiophene-2-carbaldehyde

OR346190-1G
APOSOR346190-1GEA 0 EUR
APOSOR346190-1G
5-Bromo-benzo[b]thiophene-2-carbaldehyde
5-Bromo-benzo[b]thiophene-2-carbaldehyde
Formel: C₉H₅BrOS
Molecular Weight: 241.11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11227154
CAS-Nummer: 7312-18-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR