2,2-Dimethyl-1,4-dihydro-2H-benzo[d][1,3]oxazine
Lieferant: Apollo Scientific
OR346192-1G
APOSOR346192-1GEA
0
EUR
APOSOR346192-1G
2,2-Dimethyl-1,4-dihydro-2H-benzo[d][1,3]oxazine
2,2-Dimethyl-1,4-dihydro-2H-benzo[d][1,3]oxazine
Formel:
C₁₀H₁₃NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878805 CAS-Nummer: 5226-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...