Isopropylsulphamoyl chloride

Lieferant: Apollo Scientific
Gefahr

OR346199-1G OR346199-5G
APOSOR346199-1GEA 0 EUR
APOSOR346199-1G APOSOR346199-5G
Isopropylsulphamoyl chloride
Isopropylsulphamoyl chloride
Formel: C₃H₈ClNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09753748
CAS-Nummer: 26118-67-2
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR