(Z)-2-tert-Butoxycarbonylamino-but-2-enoic acid methyl ester

Lieferant: Apollo Scientific

Synonyme: (Z)-Methyl 2-(tert-butoxycarbonylamino)but-2-enoate

OR346209-1G
APOSOR346209-1GEA 0 EUR
APOSOR346209-1G
(Z)-2-tert-Butoxycarbonylamino-but-2-enoic acid methyl ester
(Z)-2-tert-Butoxycarbonylamino-but-2-enoic acid methyl ester
Formel: C₁₀H₁₇NO₄
Molecular Weight: 215,246 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15142810
CAS-Nummer: 63658-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR