(E)-3-Bromo-2-tert-butoxycarbonylaminobut-2-enoic acid methyl ester
Lieferant: Apollo Scientific
OR346210-1G
APOSOR346210-1GEA
0
EUR
APOSOR346210-1G
(E)-3-Bromo-2-tert-butoxycarbonylaminobut-2-enoic acid methyl ester
(E)-3-Bromo-2-tert-butoxycarbonylaminobut-2-enoic acid methyl ester
Formel:
C₁₀H₁₆BrNO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD15142811 CAS-Nummer: 475288-42-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...