2,6-Dichloro-pyridine-3-sulphonyl chloride
Lieferant: Apollo Scientific
OR346214-1G
APOSOR346214-1GEA
318
EUR
APOSOR346214-1G
2,6-Dichloro-pyridine-3-sulphonyl chloride
2,6-Dichloro-pyridine-3-sulphonyl chloride
Formel:
C₅H₂Cl₃NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD06739118 CAS-Nummer: 239810-43-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...